siRNA Details
| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008868.3 | NP_032894.2 | 6 | 781 | ||
| GenBank mRNA | BC029347.1 | - | 5 | 626 | 4234596 | |
| U18119.1 | - | 5 | 607 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g2c| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008868.3 | NP_032894.2 | 6 | 781 | ||
| GenBank mRNA | BC029347.1 | - | 5 | 626 | 4234596 | |
| U18119.1 | - | 5 | 607 | |||