siRNA Details
| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293648.1 | NP_001280577.1 | 9 | 1585 | ||
| NM_019676.3 | NP_062650.1 | 9 | 1536 | |||
| GenBank mRNA | AF133125.1 | - | 9 | 1518 | ||
| AK028749.1 | - | 9 | 1537 | |||
| AK082890.1 | - | 9 | 1528 | |||
| AK197251.1 | - | 1 | 97 | |||
| BC015249.1 | - | 9 | 1490 | 4241580 | ||
| BC025798.1 | - | 9 | 1512 | 5068610 | ||
| JN635596.1 | - | 9 | 1585 | |||
| U85711.1 | - | 9 | 1530 | |||

Plcd1
Genomic Map