siRNA Details
| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293648.1 | NP_001280577.1 | 3 | 507 | ||
| NM_019676.3 | NP_062650.1 | 3 | 458 | |||
| GenBank mRNA | AF133125.1 | - | 3 | 440 | ||
| AK028749.1 | - | 3 | 459 | |||
| AK082890.1 | - | 3 | 450 | |||
| BC015249.1 | - | 3 | 412 | 4241580 | ||
| BC025798.1 | - | 3 | 434 | 5068610 | ||
| JN635596.1 | - | 3 | 507 | |||
| U85711.1 | - | 3 | 452 | |||

Plcd1
Genomic Map