siRNA Details
| Gene Symbol: Pnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008890.1 | NP_032916.1 | 3 | 788 | ||
| GenBank mRNA | BC119079.1 | - | 3 | 818 | 8733828 | |
| BC120789.1 | - | 3 | 818 | 40129712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnmt| Gene Symbol: Pnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008890.1 | NP_032916.1 | 3 | 788 | ||
| GenBank mRNA | BC119079.1 | - | 3 | 818 | 8733828 | |
| BC120789.1 | - | 3 | 818 | 40129712 | ||