siRNA Details
| Gene Symbol: Polg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017462.2 | NP_059490.2 | 21 | 3662 | ||
| GenBank mRNA | AB121698.1 | - | 20 | 3355 | ||
| AK133584.1 | - | 21 | 3588 | |||
| AK148091.1 | - | 21 | 3624 | |||
| AK162462.1 | - | 6 | 1231 | |||
| AK185475.1 | - | 2 | 76 | |||
| BC053024.1 | - | 21 | 3588 | |||
| BC061472.1 | - | 20 | 3654 | 6827238 | ||
| BC080291.1 | - | 21 | 3493 | |||
| U53584.1 | - | 21 | 3582 | |||

Polg
Genomic Map