siRNA Details
| Gene Symbol: Ppib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011149.2 | NP_035279.2 | 1 | 206 | ||
| GenBank mRNA | AK002357.1 | - | 1 | 153 | ||
| AK128977.1 | - | 1 | 156 | |||
| AK154073.1 | - | 1 | 145 | |||
| AL363449.1 | - | 1 | 247 | |||
| BC013061.1 | - | 1 | 143 | 3483267 | ||
| M60456.1 | - | 1 | 125 | |||
| X58990.1 | - | 1 | 251 | |||

Ppib
Genomic Map