siRNA Details
| Gene Symbol: Ppp2r3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163415.1 | NP_001156887.1 | 1404 | |||
| NM_011154.2 | NP_035284.2 | 1170 | ||||
| GenBank mRNA | AF050165.1 | - | 1167 | |||
| AK054122.1 | - | 1543 | ||||
| AK143212.1 | - | 1447 | ||||
| AK190355.1 | - | 198 | ||||
| BC027415.1 | - | 1147 | 4235453 | |||
| BC059852.1 | - | 1435 | 6402777 | |||
| BC095938.1 | - | 325 | ||||

Ppp2r3d
Genomic Map