siRNA Details
| Gene Symbol: Prm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008933.1 | NP_032959.1 | 1 | 162 | ||
| GenBank mRNA | AK005729.1 | - | 1 | 171 | ||
| BC049612.1 | - | 1 | 167 | 6774072 | ||
| M16456.1 | - | 1 | 65 | |||
| M27501.1 | - | 1 | 65 | |||
| X14004.1 | - | 1 | 162 | |||

Prm2
Genomic Map