siRNA Details
| Gene Symbol: Prx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019412.2 | NP_062285.1 | 5 | 399 | ||
| NM_198048.2 | NP_932165.2 | 5 | 399 | |||
| GenBank mRNA | AJ222968.1 | - | 5 | 398 | ||
| AJ222969.1 | - | 5 | 398 | |||
| AK132429.1 | - | 1 | 242 | |||
| AK173220.1 | - | 1 | 196 | |||
| BC068135.1 | - | 1 | 212 | 30354483 | ||

Prx
Genomic Map