siRNA Details
| Gene Symbol: Psma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011184.4 | NP_035314.3 | 2 | 201 | ||
| GenBank mRNA | AF055983.1 | - | 2 | 119 | ||
| AF060089.1 | - | 2 | 87 | |||
| AI303320.1 | - | 2 | 200 | |||
| AK002877.1 | - | 2 | 132 | |||
| AK165424.1 | - | 2 | 154 | |||
| AK168495.1 | - | 2 | 106 | |||
| AK169577.1 | - | 2 | 104 | |||
| AK171669.1 | - | 2 | 110 | |||
| AK172369.1 | - | 2 | 110 | |||
| BC091743.1 | - | 2 | 91 | 5030475 | ||

Psma3
Genomic Map