siRNA Details
| Gene Symbol: Psmb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013640.3 | NP_038668.2 | 5, 6 | 870 | ||
| GenBank mRNA | AK144096.1 | - | 5, 6 | 593 | ||
| BC004730.1 | - | 5, 6 | 596 | 3583052 | ||
| CT010274.1 | - | 5, 6 | 488 | |||
| D85561.1 | - | 5, 6 | 565 | |||
| U77784.1 | - | 5, 6 | 557 | |||
| Y10875.1 | - | 5, 6 | 588 | |||

Psmb10
Genomic Map