siRNA Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011186.1 | NP_035316.1 | 3 | 548 | ||
| GenBank mRNA | AB003304.1 | - | 3 | 551 | ||
| AF060091.1 | - | 3 | 548 | |||
| AK010441.1 | - | 3 | 568 | |||
| AK088367.1 | - | 3 | 300 | |||
| AK133839.1 | - | 3 | 569 | |||
| BC012246.1 | - | 3 | 556 | 4167355 | ||
| BC106144.1 | - | 3 | 551 | 4953887 | ||

Psmb5
Genomic Map