siRNA Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011186.1 | NP_035316.1 | 3 | 755 | ||
| GenBank mRNA | AB003304.1 | - | 3 | 758 | ||
| AF060091.1 | - | 3 | 755 | |||
| AK010441.1 | - | 3 | 775 | |||
| AK088367.1 | - | 3 | 507 | |||
| AK133839.1 | - | 3 | 776 | |||
| BC106144.1 | - | 3 | 758 | 4953887 | ||

Psmb5
Genomic Map