siRNA Details
| Gene Symbol: Psme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011189.1 | NP_035319.1 | 3 | 201 | ||
| GenBank mRNA | AK008514.1 | - | 3 | 176 | ||
| AK019390.1 | - | 3 | 229 | |||
| AK150708.1 | - | 3 | 245 | |||
| AK150975.1 | - | 3 | 244 | |||
| BC035549.1 | - | 2 | 359 | |||
| BC088981.1 | - | 3 | 221 | 5689048 | ||
| D87909.1 | - | 3 | 201 | |||
| U60328.1 | - | 3 | 120 | |||

Psme1
Genomic Map