siRNA Details
| Gene Symbol: Psme2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029855.1 | NP_001025026.1 | 6 | 334 | ||
| NM_011190.3 | NP_035320.1 | 6 | 434 | |||
| GenBank mRNA | AA267668.1 | - | 6 | 325 | ||
| AK012344.1 | - | 6 | 363 | |||
| AK088654.1 | - | 6 | 361 | |||
| AK143556.1 | - | 6 | 366 | |||
| BB869062.1 | - | 6 | 334 | |||
| BC005680.1 | - | 6 | 354 | 3154255 | ||
| BC057859.1 | - | 6 | 431 | 5310526 | ||
| BU563130.1 | - | 6 | 503 | |||
| CA536627.1 | - | 6 | 346 | |||
| U60329.1 | - | 6 | 303 | |||

Psme2
Genomic Map