siRNA Details
| Gene Symbol: Pspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008954.2 | NP_032980.2 | 1 | 98 | ||
| GenBank mRNA | AF040960.1 | - | 1 | 95 | ||
| BC130229.1 | - | 1 | 99 | 40141900 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pspn| Gene Symbol: Pspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008954.2 | NP_032980.2 | 1 | 98 | ||
| GenBank mRNA | AF040960.1 | - | 1 | 95 | ||
| BC130229.1 | - | 1 | 99 | 40141900 | ||