siRNA Details
| Gene Symbol: Fam195b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033231.2 | NP_001028403.1 | 3 | 222 | ||
| GenBank mRNA | AK147780.1 | - | 3 | 200 | ||
| BC002190.1 | - | 2 | 154 | |||
| BC003940.1 | - | 2 | 154 | |||
| BC025536.1 | - | 3 | 195 | |||
| BC052229.1 | - | 2 | 324 | |||
| BI987638.1 | - | 3 | 222 | |||

Fam195b
Genomic Map