siRNA Details
| Gene Symbol: Btnl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138678.2 | NP_619619.2 | 3 | 289 | ||
| GenBank mRNA | AF269232.1 | - | 3 | 289 | ||
| AK029784.1 | - | 3 | 395 | |||
| AK143638.1 | - | 3 | 439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btnl10| Gene Symbol: Btnl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138678.2 | NP_619619.2 | 3 | 289 | ||
| GenBank mRNA | AF269232.1 | - | 3 | 289 | ||
| AK029784.1 | - | 3 | 395 | |||
| AK143638.1 | - | 3 | 439 | |||