siRNA Details
| Gene Symbol: Bcas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166642.1 | NP_001160114.1 | 19 | 1941 | ||
| NM_001166643.1 | NP_001160115.1 | 18 | 1855 | |||
| NM_138681.4 | NP_619622.3 | 20 | 1986 | |||
| GenBank mRNA | AK032423.1 | - | 20 | 1937 | ||
| AK157222.1 | - | 19 | 1891 | |||
| AK171964.1 | - | 18 | 1808 | |||
| BC060690.1 | - | 19 | 1863 | |||
| BC086693.1 | - | 18 | 1820 | |||
| BC118965.2 | - | 18 | 1855 | 40097104 | ||
| BC118966.1 | - | 18 | 1837 | 40097105 | ||

Bcas3
Genomic Map