siRNA Details
| Gene Symbol: Slc25a36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138756.4 | NP_620095.1 | 3, 4 | 382 | ||
| GenBank mRNA | AK042075.1 | - | 3, 4 | 471 | ||
| AK053646.1 | - | 3, 4 | 498 | |||
| AK078779.1 | - | 3, 4 | 383 | |||
| BC008171.1 | - | 3, 4 | 374 | 3710468 | ||

Slc25a36
Genomic Map