siRNA Details
| Gene Symbol: Slc25a36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138756.4 | NP_620095.1 | 6 | 712 | ||
| GenBank mRNA | AK034423.1 | - | 2 | 248 | ||
| AK042075.1 | - | 6 | 801 | |||
| AK053646.1 | - | 6 | 828 | |||
| AK078779.1 | - | 6 | 713 | |||
| BC008171.1 | - | 6 | 704 | 3710468 | ||

Slc25a36
Genomic Map