siRNA Details
| Gene Symbol: Ptpra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163688.1 | NP_001157160.1 | 4 | 740 | ||
| NM_008980.2 | NP_033006.2 | 4 | 740 | |||
| GenBank mRNA | AF332081.1 | - | 3 | 453 | ||
| AF332082.1 | - | 3 | 453 | |||
| AJ431367.1 | - | 3 | 453 | |||
| AK010458.1 | - | 4 | 742 | |||
| AK031690.1 | - | 4 | 739 | |||
| AK165673.1 | - | 4 | 738 | |||
| BC050930.1 | - | 4 | 722 | 6811172 | ||
| BC110683.1 | - | 4 | 706 | 3965311 | ||
| M36033.1 | - | 4 | 478 | |||

Ptpra
Genomic Map