siRNA Details
| Gene Symbol: Ptprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008981.3 | NP_033007.2 | 7 | 1453 | ||
| GenBank mRNA | L09562.1 | - | 7 | 994 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprg| Gene Symbol: Ptprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008981.3 | NP_033007.2 | 7 | 1453 | ||
| GenBank mRNA | L09562.1 | - | 7 | 994 | ||