siRNA Details
| Gene Symbol: Ptprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008981.3 | NP_033007.2 | 1 | 753 | ||
| GenBank mRNA | AK053271.1 | - | 1 | 725 | ||
| BC079595.1 | - | 1 | 753 | |||
| L09562.1 | - | 1 | 294 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprg| Gene Symbol: Ptprg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008981.3 | NP_033007.2 | 1 | 753 | ||
| GenBank mRNA | AK053271.1 | - | 1 | 725 | ||
| BC079595.1 | - | 1 | 753 | |||
| L09562.1 | - | 1 | 294 | |||