siRNA Details
| Gene Symbol: Ptprj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135657.1 | NP_001129129.1 | 7 | 1331 | ||
| NM_008982.5 | NP_033008.3 | 7 | 1615 | |||
| GenBank mRNA | AK147318.1 | - | 7 | 1536 | ||
| AK147556.1 | - | 7 | 1536 | |||
| AY038891.1 | - | 7 | 1433 | |||
| AY039232.1 | - | 7 | 1433 | |||
| D45212.1 | - | 7 | 1571 | |||
| D83203.1 | - | 7 | 1332 | |||
| DQ133576.1 | - | 7 | 1346 | |||

Ptprj
Genomic Map