siRNA Details
| Gene Symbol: Ptrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008986.2 | NP_033012.1 | 2 | 1105 | ||
| GenBank mRNA | AF036249.1 | - | 2 | 1060 | ||
| BC110698.1 | - | 2 | 1087 | 30608521 | ||
| BC116701.1 | - | 2 | 1047 | 40104877 | ||
| BC117527.1 | - | 2 | 1047 | 40104876 | ||

Ptrf
Genomic Map