siRNA Details
| Gene Symbol: Igdcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008988.2 | NP_033014.1 | 2, 3 | 662 | ||
| GenBank mRNA | AF026465.1 | - | 2, 3 | 662 | ||
| AK011916.1 | - | 2, 3 | 698 | |||
| AK051027.1 | - | 2, 3 | 663 | |||
| BC053057.1 | - | 2, 3 | 639 | 5698028 | ||
| BC086481.1 | - | 2, 3 | 646 | 6505546 | ||

Igdcc3
Genomic Map