siRNA Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011223.3 | NP_035353.1 | 3 | 519 | ||
| NM_133915.3 | NP_598676.2 | 3 | 519 | |||
| GenBank mRNA | AF293882.1 | - | 3 | 444 | ||
| AF293883.1 | - | 2 | 339 | |||
| AK084243.1 | - | 1 | 184 | |||
| AK148060.1 | - | 3 | 505 | |||
| AK149933.1 | - | 3 | 484 | |||
| AK167299.1 | - | 3 | 458 | |||
| AK171226.1 | - | 3 | 508 | |||
| AK171436.1 | - | 3 | 484 | |||
| BC025493.1 | - | 3 | 477 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK089534.1 | - | 3178 | Chr. - - [-] on Build | ||

Pxn
Genomic Map