siRNA Details
| Gene Symbol: Rab33a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011228.2 | NP_035358.1 | 2 | 889 | ||
| GenBank mRNA | AB232628.1 | - | 2 | 573 | ||
| AW491145.1 | - | 1 | 291 | |||
| BC104390.1 | - | 2 | 637 | 40050293 | ||
| BC104391.1 | - | 2 | 637 | 40050296 | ||
| D83279.1 | - | 2 | 704 | |||

Rab33a
Genomic Map