siRNA Details
| Gene Symbol: Fam65b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080381.1 | NP_001073850.1 | 7 | 775 | ||
| NM_001286100.1 | NP_001273029.1 | 7 | 643 | |||
| NM_001286101.1 | NP_001273030.1 | 8 | 868 | |||
| NM_029679.2 | NP_083955.1 | 7 | 775 | |||
| NM_178658.5 | NP_848773.3 | 7 | 628 | |||
| GenBank mRNA | AK031928.1 | - | 7 | 728 | ||
| AK036452.1 | - | 8 | 848 | |||
| AK039271.1 | - | 7 | 740 | |||
| AK040887.1 | - | 7 | 608 | |||
| AK041665.1 | - | 7 | 629 | |||
| AK122269.1 | - | 7 | 707 | |||
| AK134538.1 | - | 7 | 706 | |||
| AK163880.1 | - | 7 | 711 | |||
| BC059179.1 | - | 7 | 643 | |||

Fam65b
Genomic Map