siRNA Details
| Gene Symbol: Rab3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166399.2 | NP_001159871.1 | 5 | 724 | ||
| NM_009001.6 | NP_033027.1 | 5 | 719 | |||
| GenBank mRNA | AB232587.1 | - | 4 | 534 | ||
| AK005307.1 | - | 5 | 677 | |||
| AK005362.1 | - | 5 | 681 | |||
| AK158727.1 | - | 4 | 879 | |||
| AK161975.1 | - | 5 | 688 | |||
| BC018451.1 | - | 1 | 2495 | 3489400 | ||
| BC053519.1 | - | 5 | 683 | 6467763 | ||

Rab3a
Genomic Map