siRNA Details
| Gene Symbol: Rab4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009003.3 | NP_033029.2 | 3 | 289 | ||
| GenBank mRNA | AB232591.1 | - | 3 | 149 | ||
| AK136566.1 | - | 3 | 278 | |||
| AK145452.1 | - | 3 | 272 | |||
| AK161877.1 | - | 3 | 257 | |||
| BC138508.1 | - | 3 | 202 | 8861528 | ||
| CO806247.1 | - | 305 | ||||

Rab4a
Genomic Map