siRNA Details
| Gene Symbol: Rab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293652.1 | NP_001280581.1 | 5 | 537 | ||
| NM_001293653.1 | NP_001280582.1 | 3 | 198 | |||
| NM_001293654.1 | NP_001280583.1 | 4 | 364 | |||
| NM_001293655.1 | NP_001280584.1 | 3 | 277 | |||
| NM_009005.3 | NP_033031.2 | 3 | 315 | |||
| GenBank mRNA | AB232599.1 | - | 2 | 109 | ||
| AK005004.1 | - | 3 | 277 | |||
| AK057671.1 | - | 3 | 244 | |||
| AK135659.1 | - | 4 | 365 | |||
| AK139914.1 | - | 3 | 268 | |||
| AK148117.1 | - | 3 | 308 | |||
| AK151078.1 | - | 3 | 289 | |||
| AK152666.1 | - | 5 | 512 | |||
| AK154234.1 | - | 3 | 296 | |||
| AK158971.1 | - | 3 | 199 | |||
| AK159507.1 | - | 3 | 245 | |||
| AK159928.1 | - | 3 | 216 | |||
| AK179526.1 | - | 3 | 154 | |||
| AK190348.1 | - | 3 | 154 | |||
| AK203105.1 | - | 3 | 154 | |||
| BC004597.1 | - | 3 | 233 | 3584891 | ||
| BC086793.1 | - | 3 | 231 | 6414012 | ||
| BY291181.1 | - | 3 | 316 | |||
| CT010352.1 | - | 2 | 109 | |||
| X89650.1 | - | 3 | 265 | |||

Rab7
Genomic Map