siRNA Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009012.2 | NP_033038.2 | 8 | 1398 | ||
| GenBank mRNA | AK037641.1 | - | 8 | 1399 | ||
| AK144155.1 | - | 8 | 1390 | |||
| BC058180.1 | - | 8 | 1374 | |||
| U66887.1 | - | 8 | 1376 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad50| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009012.2 | NP_033038.2 | 8 | 1398 | ||
| GenBank mRNA | AK037641.1 | - | 8 | 1399 | ||
| AK144155.1 | - | 8 | 1390 | |||
| BC058180.1 | - | 8 | 1374 | |||
| U66887.1 | - | 8 | 1376 | |||