siRNA Details
| Gene Symbol: Cnksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081047.1 | NP_001074516.1 | 21 | 1961 | ||
| GenBank mRNA | AK145910.1 | - | 21 | 1923 | ||
| BC037758.1 | - | 2 | 191 | |||
| BC113805.1 | - | 21 | 1926 | 40062208 | ||
| BC113956.1 | - | 21 | 1926 | 40062209 | ||

Cnksr1
Genomic Map