siRNA Details
| Gene Symbol: 9930104L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177573.3 | NP_808241.2 | 4, 5 | 873 | ||
| GenBank mRNA | AK158126.1 | - | 4, 5 | 848 | ||
| AK160978.1 | - | 4 | 2686 | |||
| BC099946.1 | - | 4, 5 | 753 | 30102988 | ||

9930104L06Rik
Genomic Map