siRNA Details
| Gene Symbol: Tet3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183138.2 | NP_898961.2 | 11 | 4765 | ||
| GenBank mRNA | AK084164.1 | - | 6 | 2121 | ||
| AK122274.1 | - | 1 | 68 | |||
| BC096437.1 | - | 7 | 2279 | |||
| HQ423151.1 | - | 11 | 4939 | |||
| HQ423152.1 | - | 10 | 4882 | |||
| JX946278.1 | - | 12 | 5471 | |||

Tet3
Genomic Map