siRNA Details
| Gene Symbol: Mical3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270475.1 | NP_001257404.1 | 10, 11 | 1775 | ||
| NM_153396.3 | NP_700445.2 | 10, 11 | 1741 | |||
| GenBank mRNA | AF536756.1 | - | 10, 11 | 1704 | ||
| AK048201.1 | - | 10, 11 | 1753 | |||
| AK122497.2 | - | 10, 11 | 1600 | |||
| AK132669.1 | - | 10, 11 | 1725 | |||
| BC138257.1 | - | 10, 11 | 1702 | 8861276 | ||
| BC145214.1 | - | 10, 11 | 1702 | |||

Mical3
Genomic Map