siRNA Details
Gene Symbol: Mical3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270475.1 | NP_001257404.1 | 22 | 3494 | ||
GenBank mRNA | BC043122.1 | - | 3 | 393 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mical3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270475.1 | NP_001257404.1 | 22 | 3494 | ||
GenBank mRNA | BC043122.1 | - | 3 | 393 |