siRNA Details
| Gene Symbol: Zzef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045536.2 | NP_001039001.1 | 45 | 7351 | ||
| GenBank mRNA | BC099976.1 | - | 5 | 707 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zzef1| Gene Symbol: Zzef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045536.2 | NP_001039001.1 | 45 | 7351 | ||
| GenBank mRNA | BC099976.1 | - | 5 | 707 | ||