siRNA Details
| Gene Symbol: Tmem199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199199.3 | NP_954669.2 | 5 | 570 | ||
| GenBank mRNA | AK156856.1 | - | 5 | 505 | ||
| AK158039.1 | - | 5 | 534 | |||
| BC050977.1 | - | 5 | 533 | |||
| BC064015.1 | - | 1 | 346 | |||
| BC147070.1 | - | 5 | 558 | 9056057 | ||
| BC147071.1 | - | 5 | 558 | 9056065 | ||

Tmem199
Genomic Map