siRNA Details
| Gene Symbol: Tmem199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199199.3 | NP_954669.2 | 6 | 628 | ||
| GenBank mRNA | AK156856.1 | - | 6 | 563 | ||
| AK158039.1 | - | 6 | 592 | |||
| AK160676.1 | - | 1 | 34 | |||
| BC050977.1 | - | 6 | 591 | |||
| BC064015.1 | - | 2 | 404 | |||
| BC147070.1 | - | 6 | 616 | 9056057 | ||
| BC147071.1 | - | 6 | 616 | 9056065 | ||

Tmem199
Genomic Map