siRNA Details
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009029.2 | NP_033055.2 | 23 | 2531 | ||
GenBank mRNA | AK132875.1 | - | 12 | 1277 | ||
AK141028.1 | - | 23 | 2563 | |||
AK146467.1 | - | 18 | 1862 | |||
BC096525.1 | - | 23 | 2531 | 6832808 | ||
DQ400415.1 | - | 23 | 2381 | |||
M26391.1 | - | 23 | 2479 |
