siRNA Details
| Gene Symbol: Robo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164767.1 | NP_001158239.1 | 10 | 1773 | ||
| GenBank mRNA | AF060570.1 | - | 9 | 1759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Robo3| Gene Symbol: Robo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164767.1 | NP_001158239.1 | 10 | 1773 | ||
| GenBank mRNA | AF060570.1 | - | 9 | 1759 | ||