siRNA Details
| Gene Symbol: Rbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009034.4 | NP_033060.3 | 2 | 287 | ||
| GenBank mRNA | AK003312.1 | - | 2 | 272 | ||
| BC125551.1 | - | 2 | 256 | 40130066 | ||
| BC125553.1 | - | 2 | 256 | 40130068 | ||
| X74154.1 | - | 2 | 271 | |||

Rbp2
Genomic Map