siRNA Details
| Gene Symbol: Rbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009034.4 | NP_033060.3 | 2, 3 | 321 | ||
| GenBank mRNA | AK003312.1 | - | 2, 3 | 306 | ||
| BC125551.1 | - | 2, 3 | 290 | 40130066 | ||
| BC125553.1 | - | 2, 3 | 290 | 40130068 | ||
| X74154.1 | - | 2, 3 | 305 | |||

Rbp2
Genomic Map