siRNA Details
| Gene Symbol: Rfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011258.2 | NP_035388.2 | 15 | 2078 | ||
| GenBank mRNA | BC085173.1 | - | 15 | 2055 | 30549691 | |
| BC085174.1 | - | 15 | 2045 | |||
| U01222.1 | - | 15 | 2046 | |||
| U07157.1 | - | 15 | 2041 | |||
| U36441.1 | - | 15 | 2052 | |||
| X72711.1 | - | 15 | 2044 | |||

Rfc1
Genomic Map