siRNA Details
| Gene Symbol: Rest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011263.2 | NP_035393.2 | 4 | 1304 | ||
| GenBank mRNA | AB024496.1 | - | 3 | 1000 | ||
| AB221589.1 | - | 3 | 999 | |||
| AK004945.2 | - | 4 | 1307 | |||
| AK156514.1 | - | 4 | 1290 | |||
| BC096434.1 | - | 4 | 1227 | |||
| BC127065.1 | - | 3 | 1000 | 40110015 | ||

Rest
Genomic Map