siRNA Details
| Gene Symbol: Rgs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009063.3 | NP_033089.2 | 5 | 481 | ||
| GenBank mRNA | AK004165.1 | - | 5 | 482 | ||
| AK028765.1 | - | 484 | ||||
| AK030300.1 | - | 483 | ||||
| AK044096.1 | - | 5 | 482 | |||
| AK054098.1 | - | 5 | 482 | |||
| BC037683.1 | - | 5 | 468 | 3962083 | ||
| U67188.1 | - | 438 | ||||

Rgs5
Genomic Map