siRNA Details
| Gene Symbol: Ring1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009066.3 | NP_033092.3 | 7 | 1363 | ||
| GenBank mRNA | AK154962.1 | - | 7 | 1324 | ||
| AK155509.1 | - | 7 | 1339 | |||
| BC009070.1 | - | 7 | 1326 | 2651194 | ||
| BC082771.1 | - | 3 | 2173 | |||
| BU505852.1 | - | 2 | 130 | |||
| BU918502.1 | - | 3 | 406 | |||
| CT010219.1 | - | 5 | 1053 | |||
| CX219100.1 | - | 2 | 291 | |||
| KJ650216.1 | - | 6 | 1140 | |||
| Y12881.1 | - | 5 | 1059 | |||

Ring1
Genomic Map